Learn More
R&D Systems™ Recombinant S. cerevisiae PPA1 Protein
Description
Order Info
The product is shipped with polar packs. Upon receipt, store it immediately at the recommended storage temperatures.
Specifications
Specifications
| Accession Number | P00817 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, DTT and MgCl. |
| Gene ID (Entrez) | 5464 |
| Molecular Weight (g/mol) | M.W. (Observed): 36 kDa, reducing conditions; M.W. (theoretical): 33 kDa |
| Quantity | 50 μg |
| Source | E. coli-derived yeast Inorganic Pyrophosphatase/PPA1 protein Thr2-Val287, with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <0.01 EU / 1 μg of the protein by the LAL method. |
| Gene Symbol | IPP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.