Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant S. pneumoniae beta(1-4)-Galactosidase

Catalog No. 5549GH100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5549GH100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 5549GH100 Supplier R&D Systems™ Supplier No. 5549GH100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to cleave a fluorogenic substrate, 4-Methylumbelliferyl-beta-D-galactopyranoside. The specific activity is >600 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number YP_815516
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Molecular Weight (g/mol) M.W. (Observed): 62 kDa, reducing conditions; M.W. (theoretical): 70 kDa
Name beta (1-4)-Galactosidase
Quantity 100 μg
Source E. coli-derived s. pneumoniae beta (1-4)-Galactosidase protein Thr2-Leu595, with an N-terminal Met and a 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein beta 1-4)-Galactosidase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.