Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Saccharomyces cerevisiae GLC8 His Protein
SDP

Catalog No. NBC118408 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBC118408 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBC118408 Supplier Novus Biologicals™ Supplier No. NBC118408
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-229 of Saccharomyces cerevisiae GLC8 The Recombinant Yeast GLC8 Protein is derived from E. coli. The Recombinant Yeast GLC8 Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 1 mg/mL
For Use With (Application) SDS-PAGE
Formulation 20 mM Tris-HCl buffer (pH 8.0), 1 mM DTT, 10% glycerol
Gene ID (Entrez) 855359
Molecular Weight (g/mol) M.W. (theoretical): 30.7 kDa
Name GLC8 Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen 1-229aa, Saccharomyces cerevisiae, His-tagged, Recombinant, E.coli
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Regulatory Status RUO
Gene Symbol GLC8
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Research Category Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Protein Saccharomyces cerevisiae GLC8
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.