Learn More
R&D Systems™ Recombinant SARS-CoV-2 B.1.617.1 S Alexa Fluor™ 488 Protein
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 43740568 |
| Molecular Weight (g/mol) | M.W.-theroretical: 138 kDa, M.W.-observed: 139-167 kDa, under reducing conditions. |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein Recombinant SARS-CoV-2 Spike (Val16-Lys1211)(Gly142Asp, Glu154Lys, Leu452Arg, Glu484Gln, Asp614Gly, Pro681Arg, Gln1071His)(Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro) (c-terminus) Accession # YP_009724390.1 GCN4-IZ 6-His tag N-terminus C-terminμs |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.