Learn More
R&D Systems™ Recombinant SARS-CoV-2 D614G Spike S1 Subunit His Protein CF
Shop All R&D Systems Products

Description
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| Conjugate | Unconjugated |
| For Use With (Application) | Bioactivity |
| Quantity | 100 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Synonym | SARS-CoV-2 |
| Target | SARS-CoV-2 |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.