Learn More
Specifications
Specifications
| Formulation | 0.22 μm filtered solution in PBS, pH 7.4. Normally trehalose is added as protectant before lyophilization. |
| Gene ID (Entrez) | 43740570 |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 9.6 kDa |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C to -70°C. Avoid freeze/thaw cycles. |
| Gene Alias | 2019-nCoV Envelope protein, COVID-19 Envelope protein, E Protein, Envelope, Human Coronavirus Envelope protein, SARSCoV2, SARS-CoV-2 E protein, SARSCoV2 Envelope protein, SARS-CoV-2 Envelope protein, Severe acute respiratory syndrome 2 Envelope Protein |
| Conjugate | Unconjugated |
| Species | SARS-CoV-2 |
| Recombinant | Recombinant |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
