Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 NSP13 His (C-Term) Protein
SDP

Catalog No. NB151294 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151294 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151294 Supplier Novus Biologicals™ Supplier No. NBP307929
Add to Cart
Add to Cart

Recombinant Protein

Specifications

Formulation 0.2 um filtered solution of 20 mM Tris-HCl, 150 mM NaCl, 10% Glycerol, pH 8.5.
Quantity 0.025 mg
Storage Requirements Store at -20 to -70 C. Avoid freeze-thaw cycles.
Gene Symbol ORF1ab
Purity or Quality Grade >80%, by SDS-PAGE
Protein nsp13
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.