Learn More
Specifications
Specifications
| Accession Number | YP_009725311.1 (NSP16) and YP_009725306.1 (NSP10) |
| Conjugate | Unconjugated |
| Product Line | R&D Systems |
| Quantity | 100 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Target | SARS-CoV-2 |
| Buffer | Supplied as a 0.2 μm filtered solution in Tris, NaCl, TCEP and Glycerol. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
