Learn More
R&D Systems™ Recombinant SARS-CoV-2 NSP8 His-tag Protein
Specifications
Specifications
| Accession Number | YP_009725304.1 |
| Conjugate | Unconjugated |
| Quantity | 100 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Synonym | COVID-19 |
| Target | SARS-CoV-2 |
| Buffer | Supplied as a 0.2 μm filtered solution in HEPES and NaCl. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.