Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant SARS-CoV-2 S GCN4-IZ Alexa Fluor™ 647 Protein

Catalog No. AFR10796020
Change view
Click to view available options
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AFR10796020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR10796020 Supplier R&D Systems™ Supplier No. AFR10796020
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

B.1.1.7 Alpha Variant (UK) His-tag

Measured by flow cytometry for its ability to bind HEK293 human embryonic kidney cells transfected with human ACE-2 at 0.25-1.00 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number YP_009724390.1
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 43740568
Molecular Weight (g/mol) M.W.-theroretical: 138 kDa, M.W.-observed: 140-170 kDa, under reducing conditions.
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike protein Val16-Lys1211(His69del, Val70del, Tyr145del, Asn501Tyr, Ala570Asp, Asp614Gly, Pro681His, Thr716Ile, Ser982Ala, Asp1118His)(Arg682Ser, Arg685Ser, Lys986Pro, Val987Pro), with a C-terminal GCN4-IZ & 6-His tag
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias 2019-nCoV S Protein, 2019-nCoV Spike, COVID-19 Spike, E2, Human coronavirus spike glycoprotein, Peplomer protein, S glycoprotein, S Protein, SARS-CoV-2, SARS-COV-2 S protein, SARS-COV-2 Spike glycoprotein, SARSCOV2 Spike protein, Severe Acute Respiratory Syndrome Coronavirus 2 Spike Protein, Spike glycoprotein, surface glycoprotein
Conjugate Alexa Fluor 647
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein SARS-CoV-2 Spike
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.