Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Recombinant SARS-CoV-2 S1+S2 ECD Protein (His tag, pre-fusion state)

Catalog No. MP08L100010 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP08L100010 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MP08L100010 Supplier MP Biomedicals™ Supplier No. 08L100010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant SARS-CoV-2 S1+S2 ECD Protein is ultra pure and possesses a high biological activity, which can be apllied to a variety of scientific experiments related to SARS-CoV-2 research.

Recombinant SARS-CoV-2 S1+S2 ECD (S-ECD) Protein is produced by HEK293 cells expression system. The target protein is expressed with sequence (Val11-Gln1208) of SARS-CoV-2 S1+S2 ECD (S-ECD) (Accession #YP_009724390.1) fused with a 6×His tag at the C-terminus. It is the wildtype sequence in pre-fusion state without any mutation.

  • Eukaryotic (Expi293) system-expressed protein
  • High bioactivity determined with ELISA
  • High purity determined with SDS-PAGE
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mg/mL if dissolve in 100 μL of ddH20 or biological buffer
For Use With (Application) Binding Assay, ELISA, Immune Blocking, Immunoprecipitation, SDS-PAGE, Surface Plasmon Resonance (SPR), Western Blot
Formulation Lyophilized from a 0.22 μm filtered solution of PBS, pH 7.3 to 7.4
Gene ID (Entrez) 43740568
Preparation Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Endotoxin Concentration < 0.1 EU/μg of the protein by LAL method
Gene Alias S glycoprotein; S protein; Spike glycoprotein
Conjugate Unconjugated
Species SARS-CoV-2
Protein Tag C-6xHis
Content And Storage -20°C
Target Species Named Human, Mouse, Rat
Expression System HEK293 cells
Color White
Purity or Quality Grade > 97% by SDS-PAGE
Protein SARS-CoV-2 S1+S2 ECD(S-ECD)
Solubility Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water. Addition of a carrier protein (such as 0.05-0.1% BSA) is recommended for long-term storage
Show More Show Less

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.