Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV-2 Spike S1 (RBD) His (C-Term) Protein
SDP

Catalog No. NB025786 Shop All R&D Systems Products
Change view
Click to view available options
:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NB025786 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB025786 Supplier Novus Biologicals™ Supplier No. NBP290982
Only null left
Add to Cart
Add to Cart

Measured by its binding ability in a functional ELISA.

Measured by its binding ability in a functional ELISA. Immobilized Recombinant 2019-nCoV RBD-6His at 2ug/mL (100 uL/well) can bind Recombinant Human ACE2 with a linear range of 8-80 ng/mL.

Specifications

Concentration LYOPH
For Use With (Application) ELISA, SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution PBS (pH 7.4), 5% trehalose and 5% mannitol
Gene ID (Entrez) 43740568
Quantity 100 μg
Storage Requirements Store at -20 to -70 C. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/ug of the protein by LAL method.
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water.
Research Category Virology Bacteria and Parasites
Purity or Quality Grade >95%, by SDS-PAGE
Protein SARS-CoV-2 Spike S1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.