Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S1 Subunit Biotin Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| Conjugate | Biotin |
| For Use With (Application) | Bioactivity |
| Quantity | 50 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Synonym | SARS-CoV-2 |
| Target | SARS-CoV-2 |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.