Learn More
Specifications
Specifications
| Formulation | 0.2 uM filtered solution of PBS, pH 7.4 |
| Gene ID (Entrez) | 43740568 |
| Molecular Weight (g/mol) | 8.4 kDa |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20°C. |
| Gene Alias | COVID-19 Spike S2 HR1, S2 Protein Heptad Repeat 1, SARS-CoV-2 Spike S2 Heptad Repeat 1, Spike glycoprotein Subunit 2 HR1, Spike S2 4,3 hydrophobic repeat region 1 |
| Gene Symbol | S |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Spike S2 HR1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
