Learn More
R&D Systems™ Recombinant SARS-CoV-2 Spike S2 Subunit His-tag Protein
Description
Specifications
Specifications
| Accession Number | YP_009724390.1 |
| Conjugate | Unconjugated |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 43740568 |
| Quantity | 100 μg |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Synonym | COVID-19, S2 protein, SARS-CoV-2, SARS-CoV-2 Spike, Spike glycoprotein Subunit 2, spike protein, Spike protein S2, structural protein, surface glycoprotein |
| Target | SARS-CoV-2 |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.