Learn More
Specifications
Specifications
| For Use With (Application) | ELISA, Western Blot |
| Formulation | 50 mM Tris-HCl, 60 mM NaCl, 50% Glycerol |
| Quantity | 0.05 mg |
| Storage Requirements | Store at -20 to -70 C. Avoid freeze-thaw cycles. |
| Gene Symbol | M |
| Product Line | Novus |
| Purity or Quality Grade | >95%, by 10% PAGE (coomassie staining). Purified by proprietary chromatographic technique. |
| Protein | Membrane |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
