Learn More
R&D Systems™ Recombinant SARS-CoV Nucleocapsid His-tag Protein
Description
Specifications
Specifications
| Accession Number | NP_828858.1 |
| For Use With (Application) | Flow Cytometry |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS and EDTA with Trehalose. |
| Molecular Weight (g/mol) | M.W.-observed: 41-55 kDa, under reducing conditions, M.W.-theroretical: 47 kDa |
| Quantity | 100 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived sars-cov Nucleocapsid protein Met1-Ala422, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 1 mg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.