Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant SARS-CoV Spike S2 (1051-1076, 1121-1154, 1162-1190 aa) Protein
SDP

Catalog No. NB151310 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151310 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB151310 Supplier Novus Biologicals™ Supplier No. NBP307945
Only null left
Add to Cart
Add to Cart

Recombinant Protein

Specifications

For Use With (Application) ELISA, Western Blot
Formulation 25 mM Tris-HCl, 0.4% Sarcosyl, 0.25% Triton X-100, 50% glycerol
Quantity 0.05 mg
Storage Requirements Store at -20 to -70 C. Avoid freeze-thaw cycles.
Purity or Quality Grade >95%, by 10% PAGE (coomassie staining). Purified by proprietary chromatographic technique.
Protein Spike S2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.