Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant SARS-CoV2 BA.4/BA.5 RBD Alexa Fluor™ 647 Protein

Catalog No. AFR11229020
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AFR11229020 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AFR11229020 Supplier R&D Systems™ Supplier No. AFR11229020
Add to Cart
Add to Cart
This item is not returnable. View return policy

His-tag, Omnicron Variant

Measured by flow cytometry for its ability to bind HEK293 human embryonic kidney cells transfected with human ACE-2 at 0.250-1.00 μg/mL (100 μL/well). Please Note: Optimal dilutions should be determined by each laboratory for each application.

Specifications

Accession Number YP_009724390.1
Formulation Supplied as a 0.2 μm filtered solution in PBS with BSA as a carrier protein.
Molecular Weight (g/mol) M.W.-observed: 33-39 kDa, under reducing conditions., M.W.-theroretical: 26 kDa
Quantity 20 μg
Source Human embryonic kidney cell, HEK293-derived sars-cov-2 Spike RBD protein Arg319-Phe541 (Gly339Asp, Ser371Phe, Ser373Pro, Ser375Phe, Thr376Ala, Asp405Asn, Arg408Ser, Lys417Asn, Asn440Lys, Leu452Arg, Ser477Asn, Thr478Lys, Glu484Ala, Phe486Val, Gln498Arg, Asn501Tyr, Tyr505His), with a C-terminal 6-His tag
Storage Requirements Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Conjugate Alexa Fluor 647
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Spike RBD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.