Learn More
Abcam Recombinant Toxoplasma gondii Protein, E. coli, >95% Purity, 100 ug
Description
- Protein Type: Recombinant
- Source: Escherichia coli
- Fragment: 234 to 306 aa
- Purity: >95%
- Applications: ELISA, WB, Flow Cyt
- Formulation: Liquid, pH 7.2 to 7.6, 50% Glycerol, 9% Urea, 0.395% Tris HCl, 0.0292% EDTA, and 0.01% Sodium azide
- Storage Conditions: Store at -20°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Toxoplasma gondii |
| Shipping Condition | Blue Ice |
| Reactivity | Toxoplasma gondii |
| Label or Dye | Unconjugated |
| Form | Liquid |
| Alternate Part Number | P30_TOXGO |
| Buffer | 50% glycerol, 9% urea, 0.395% Tris-HCl, 0.0292% EDTA, 0.01% sodium azide, pH7.4 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Formulation | 50% Glycerol, 9% Urea, 0.395% Tris HCl, 0.0292% EDTA, 0.01% Sodium azide, pH 7.2 to 7.6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.