Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Viral 136R/Y136 Protein
Click to view available options
:
25μg
Description
The Recombinant Viral 136R/Y136 Protein is derived from CHO. The Recombinant Viral 136R/Y136 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
For Use With (Application) | Bioactivity |
Molecular Weight (g/mol) | Observed M.W.: 88-120kDa, reducing conditions, Theoretical M.W.: 41kDa |
Quantity | 25μg |
Source | Chinese Hamster Ovary cell line,CHO-derived viral 136R protein, (N-terminus) Viral 136R/Y136 (Tyr17-Lys351) Accession &Num; Q9DHH7 HPGGGSGGGSGGGS HHHHHH (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
Common Name | 136R |
Biological Activity | Measured by its ability to inhibit Type-I and Type-III IFN-mediated anti-viral activity. Huang, J. et al. (2007) PANS 104:9822. The ED50 for this effect is typically 2-12ng/mL in the presence of 20 pg/mL Recombinant Human IFN-beta. |
Conjugate | Unconjugated |
Species | Virus |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction