Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | Observed M.W.: 88-120kDa, reducing conditions, Theoretical M.W.: 41kDa |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived viral 136R protein, (N-terminus) Viral 136R/Y136 (Tyr17-Lys351) Accession &Num; Q9DHH7 HPGGGSGGGSGGGS HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Common Name | 136R |
| Biological Activity | Measured by its ability to inhibit Type-I and Type-III IFN-mediated anti-viral activity. Huang, J. et al. (2007) PANS 104:9822. The ED50 for this effect is typically 2-12ng/mL in the presence of 20 pg/mL Recombinant Human IFN-beta. |
| Conjugate | Unconjugated |
| Species | Virus |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
