Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Viral Dengue Virus 4 NS1 Protein
SDP

Catalog No. 9444DG100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94-44D-G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 94-44D-G100 Supplier R&D Systems™ Supplier No. 9444DG100
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Viral Dengue Virus 4 NS1 Protein CF is a non-structural protein that is a viral glycoprotein.

The Recombinant Viral Dengue Virus 4 NS1 Protein is derived from HEK293.

Specifications

Accession Number P09866
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 40kDa
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived viral Dengue Virus 4 NS1 protein Asp775-Ala1126,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias DEN-4, Dengue NS1, Dengue pan, Dengue virus Serotype 4, Dengue virus Type 4, DENV4, DENV4-NS1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Virus
Reconstitution Reconstitute at 500μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.