Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein

Catalog No. 9450ZK100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
94-50Z-K100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 94-50Z-K100 Supplier R&D Systems™ Supplier No. 9450ZK100
Add to Cart
Add to Cart
This item is not returnable. View return policy

R&D Systems™ Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein CF is a non-structural protein 1 that is a viral glycoprotein.

The Recombinant Viral Zika Virus (H/PF/2013) NS1 Protein is derived from HEK293.

Specifications

Accession Number AHZ13508
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol) 41kDa
Quantity 100 μg
Source Human embryonic kidney cell,HEK293-derived viral Zika Virus (H/PF/2013) NS1 protein Val796 & Ser799-Ser1148,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Product Type Recombinant Protein
Conjugate Unconjugated
Species Virus
Reconstitution Reconstitute at 250μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.