Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals Recombinant Virus Dengue Virus 4 Envelope Dominica/814669/1981 His (C-Term) Protein
SDP

Catalog No. p-7229493 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159108 0.1 mg
NB159109 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159108 Supplier Novus Biologicals Supplier No. NBP3148150.1MG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl, 110mM sodium chloride, pH7.8.
Quantity 0.1 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Dengue Virus 4 Envelope
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.