Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Virus Dengue Virus 4 Envelope Dominica/814669/1981 His (C-Term) Protein
SDP

Catalog No. NB159109 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
0.1 mg
0.5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB159109 0.5 mg
NB159108 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB159109 Supplier R&D Systems™ Supplier No. NBP3148150.5MG

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

Specifications

For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl, 110mM sodium chloride, pH7.8.
Quantity 0.5 mg
Storage Requirements Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Product Line Novus
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Dengue Virus 4 Envelope
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.