Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Virus Hepatitis E Virus Capsid Fc Protein
Shop All R&D Systems Products

Click to view available options
Quantity:
0.1 mg
0.5 mg
Specifications
Specifications
For Use With (Application) | SDS-PAGE |
Formulation | DPBS pH7.4 |
Molecular Weight (g/mol) | theroretical M.W.: 96 kDa |
Quantity | 0.1 mg |
Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Gene Alias | ORF2, pORF2, Secreted protein ORF2 |
Purity or Quality Grade | >95% |
Protein | Hepatitis E Virus Capsid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction