Learn More
Novus Biologicals Recombinant Virus Zika Virus NS1 Suriname Z1106033 His (C-Term) Protein
Shop All R&D Systems Products

Specifications
Specifications
| Formulation | DPBS pH 7.4 |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
| Gene Alias | polyprotein, partial |
| Purity or Quality Grade | >90% SDS-PAGE |
| Protein | Zika Virus NS1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.