Learn More
R&D Systems™ Reconstitution Buffer 1 (PBS)
SDP
Catalog No. RB01
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
RB01 2 mL
1 options
This item is not returnable. View return policy
Catalog No. RB01 Supplier R&D Systems™ Supplier No. RB01
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Phosphate-Buffered Saline
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
Specifications
Quantity 2 mL