Learn More
R&D Systems™ Reconstitution Buffer 2 (BSA/PBS)
SDP
Catalog No. RB02 Shop All Novus Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
RB02 2 mL
1 options
This item is not returnable. View return policy
Catalog No. RB02 Supplier R&D Systems™ Supplier No. RB02
Add to Cart
Add to Cart
This item is not returnable. View return policy
0.1% Bovine Serum Albumin in PBS
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
Specifications
Product Line R&D Systems
Quantity 2 mL