Learn More
R&D Systems™ Reconstitution Buffer 6 (BSA/Acetic Acid)
SDP
Catalog No. RB06 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
RB06 2 mL
1 options
This item is not returnable. View return policy
Catalog No. RB06 Supplier R&D Systems™ Supplier No. RB06
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
0.1% Bovine Serum Albumin in 100 mM Acetic Acid
This product is intended as a companion product for reconstitution of selected lyophilized antibody or protein to maximize recovery and activity. Refer to the specific antibody or protein product data sheet for the recommended reconstitution concentrations.
Specifications
Product Line R&D Systems
Quantity 2 mL