Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innocal™ Recorder and Datalogger Calibrations

Catalog No. 1509449 Shop All Oakton Products
Change view
Click to view available options
Product Type:
Electrical (X-Y Recorder)
pH Meter, Handheld/Benchtop
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
15-094-49 Electrical (X-Y Recorder)
15-094-28 pH Meter, Handheld/Benchtop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 15-094-49 Supplier Innocal™ Supplier No. WD1710010
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

If your experiment or process relies on recorded measurements, make sure these instruments remain precise through regular InnoCal™ NIST-Traceable Calibrations for Recorders and Data Loggers

  • Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal.
  • The results stated in our reports relate only to the items tested or calibrated.
  • Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005.

Specifications

For Use With (Equipment) X-Y Recorders
Certifications/Compliance NIST-Traceable
Description Calibrated per manufacturer's procedure
Duration 12 Months
Calibration Type Electrical (X-Y Recorder)
Product Type Electrical (X-Y Recorder)
For Use With (Application) Calibration
Includes Calibrated per manufacturer's procedure
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.