Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Recoverin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7087696 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6002 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6002 Supplier Proteintech Group Inc Supplier No. 100731AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcium binding proteins are implicated as being a helper molecule in aberrant tissue growth of tumors. Recoverin is a neurologically important calcium binding protein, involved in tumor pathology. It is conserved across species and there are indications of several isoforms of this molecule. Recoverin antibodies are a useful tool for distinguishing between pineal gland-derived tissue and the surrounding tissue of the brain. They can also be used to accurately determine maligt tissue in this location of the brain. In pineal tumor tissue, the normal tissue staining pattern is lost, indicating the tissue′s loss of this protein.

Specifications

Antigen Recoverin
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 0.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RCVRN
Gene Accession No. P34057, P35243
Gene Alias Protein CAR, RCV1, RCVRN, recoverin
Gene Symbols RCVRN
Host Species Rabbit
Immunogen Recoverin Fusion Protein Ag0119
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140936, 19674, 5957
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.