Learn More
Alkali Scientific ASI™ 1" wide x 60yd Color Coding Labeling Tape, 2160 Labels per Roll, 3/Rolls, Maroon
SDP
Supplier:  Alkali Scientific T100MAROON
Encompass_Preferred
Color Labeling Tape is our another popular item, which is offered in an assortment of colors. This tape is beneficial in any laboratory, medical, or educational setting. From labeling test tubes to laboratory wraps, our brand color coding tape is an essential! Strong and Durable – oil, acid, and water resistant Withstands temperatures from -23º C to +121ºC (-10º F to +250ºF) Can be written on with pen, pencil, or permanent marker without smearing Available in 27 different colors and 1/2”, 3/4”, 1”, 1 1/2”, and 2” widths Labels Per Case: 2160", 3 rolls/case
Catalog No. 50-302-8290
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.