Learn More
MSC RECTANGULAR INSPECTION MIRROR
Supplier:  MSC 67735217
Encompass
PROTO15-1/2″ OAL, Rectangular Inspection Mirror3-1/2″ Long, 2-1/8″ Wide, 11-1/4″ Collapsed Length, Telescoping
Catalog No. NC2124450
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.