Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Rectangular Roof Leak Diverter Tarp Clear
SDP

Catalog No. 18366645 Shop All New Pig Corporation Products
Change view
Click to view available options
Material:
Polyester-blended PVC
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
18-366-645 Polyester-blended PVC
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-645 Supplier PIG™ Supplier No. TLS672CL
Add to Cart
Add to Cart

Rectangular leak diverter trap that quickly installs to channel away roof or ceiling leaks before it damages inventory and machinery or causes a slip hazard.

  • 3 ft. x 10 ft. leak diverter tarp
  • Keeps rain and snow from coming inside
  • Heavy-duty vinyl-coated tarp resists punctures, UV and mildew and lasts multiple deployments
  • Pinched corners create a funnel to channel liquids to a drain or collection container without overflowing or sagging, engineered with leak-proof seams and connections
  • Brass grommets resist tearing and make installation easy, multiple hanging points add stability and preserve funnel shape
  • Extra-large drain opening and debris cage prevents clogs from crumbled ceiling tiles, drywall and roofing material
  • Accepts a standard 3/4 in. hose connection (not included)
  • Flow rate of 3.33 gal./min. when used with a 1/2 in. ID hose
  • Collection container, hose and hanging straps sold separately

Specifications

Color Clear
Material Polyester-blended PVC
Type Roof Leak Diverter Tarp
Length (English) 10 ft.
Length (Metric) 3 m
Width (English) 3 ft.
Width (Metric) 91 cm
Weight (English) 2.27 lb.
Weight (Metric) 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.