Learn More
Amsbio LLC Rectum adenocarcinoma tissue array/ including TNM and pathology grade/ 75 cases/150 cores
Supplier:  Amsbio LLC RE1501
Encompass
Rectum adenocarcinoma tissue microarray/ containing 54 cases of rectum adenocarcinoma/ 8 rectum mucinous adenocarcinoma/ 5 rectum signet-ring cell carcinoma/ 2 rectum squamous cell carcinoma and 1 rectum atypical carcinoid/ plus 5 normal rectum tissue/ duplicate cores per case
Catalog No. NC3007575