Learn More
MilliporeSigma™ ReCycler™ Dispensing Hose Assembly
Catalog No. M6911222 Shop All MilliporeSigma Products
Change view
Click to view available options
For Use With:
Most Closed EMD ReCycler Systems
Catalog No. For Use With
M6911222 Most Closed EMD ReCycler Systems
1 options
Catalog No. M6911222 Supplier MilliporeSigma™ Supplier No. 6911222
Add to Cart
Add to Cart
Use with most closed ReCycler™ assemblies. MilliporeSigma™ ReCycler™ Dispensing Hose Assembly is a stainless steel hose assembly with a thumb trigger and Swagelok™ quick-connect coupling.
Dispenser Hose/Handheld Applications New, safer thumb trigger. Durable all stainless steel design. Easy Swagelok™ quick connect coupling. For use on most closed EMD ReCycler™ systems.
TRUSTED_SUSTAINABILITY
Specifications
Description Dispensing Hose Assembly with thumb trigger and Swagelok quick connect coupling
For Use With Most Closed EMD ReCycler Systems
Material Stainless Steel