Learn More
Description
These collectors are made from a minimum 70% recycled material and are intended for Resource Conservation and Recovery Act (RCRA) solid hazardous waste. As an added feature, each RCRA collector comes with a liquid-absorbent material preattached to the inside bottom of the collector.
Specifications
Specifications
| Capacity (Metric) | 34.1 L |
| Depth (English) | 28.6 in. |
| Depth (Metric) | 72.6 cm |
| Capacity (English) | 9 gal. |
| Color | Gray |
| Height (Metric) | 59.6 cm |
| Sterility | Non-sterile |
| Width (Metric) | 41.4 cm |
| Includes | Liquid-absorbent material |
| For Use With (Application) | Design that reduces the risk of collectors tipping over |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.