Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Red Banana/Alligator Cord
SDPSCI_ED

Catalog No. S81071 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (English):
12 in.
24 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S81071 12 in.
S81062 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S81071 Supplier United Scientific Supplier No. WAB012R
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Banana/Alligator Cord for use with multiple electricity demonstrations.

  • Flexible, 18 gauge (SWG) stranded wire cord with a stackable banana plug at one end and an alligator clip at the other end
  • Alligator clip has a protective vinyl sheath
  • 12 in., 24 in., or 36 in. lengths
  • Prop 65 Warning Label

Specifications

Length (English) 12 in.
For Use With (Application) Electricity Demonstration
Product Type Wires
Color Red
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.