Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red Blood Cells, Sheep, MP Biomedicals

Catalog No. ICN55876 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN55876 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN55876 Supplier MP Biomedicals, Inc Supplier No. 0855876

missing translation for 'validMainframeMsg'

Washed, preserved red blood cells in a 10% suspension

TRUSTED_SUSTAINABILITY

Specifications

Buffer Phosphate Buffered Saline
Certifications/Compliance USDA
Content And Storage 2° to 8°C
Quantity 20 mL
Source Sheep
For Use With (Application) Immunoassays (rosetting), Hemolytic and Hemagglutination Assays and Assays of Heme Inhibition
Preservative Borate Buffer, Sodium Acetate
Test Methods USDA Inspected
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.