Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Red/Blue Bar Magnets, Pair
SDPSCI_ED

Catalog No. s66005 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S66005 5+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S66005 Supplier United Scientific™ Supplier No. MPC080
Only null left
Add to Cart
Add to Cart

Demonstrate the forces of attraction and repulsion with this pair of Red/Blue Bar Magnets.

  • Provide an easy way to teach magnetic attraction and repulsion.
  • Plastic covered.
  • Dimensions: 3.2 x 0.9 x 0.4 in. (80 x 23 x 10 mm)

Specifications

Material Alnico
Shape Rectangular
For Use With (Application) Demonstrate the forces of attraction and repulsion
Grade 5+
Color Red/Blue
Product Type Bar Magnet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.