Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Red Buffer Pad

3M™ Red Buffer Pads are especially designed for spray buffing. Cleans when damp; buffs when dry. Quickly cleans and removes scuff marks and enhances floor appearance. Works great on automatic floor scrubbers for light duty cleaning.

$27.79 - $100.98

Specifications

Product Type Cleaning Pad
Color Red
For Use With (Application) Buffing, Cleaning
For Use With (Equipment) Rotary or Automatic Floor Machines
Material Non-Woven Polyester Fiber
View More Specs

Products 9
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Price Quantity & Availability  
17-623-808
View Documents
3M™
7000000676
305 mm
Case of 5 for $27.79
Only null left
Add to Cart
 
17-623-810
View Documents
3M™
7000000678
356 mm
Case of 5 for $35.70
Only null left
Add to Cart
 
17-623-811
View Documents
3M™
7000000679
380 mm
Case of 5 for $39.02
Only null left
Add to Cart
 
17-623-812
View Documents
3M™
7000000680
406 mm
Case of 5 for $41.82
Only null left
Add to Cart
 
17-623-803
View Documents
3M™
7000000662
432 mm
Case of 5 for $46.41
Only null left
Add to Cart
 
17-623-813
View Documents
3M™
7000000681
460 mm
Case of 5 for $49.73
Only null left
Add to Cart
 
17-623-814
View Documents
3M™
7000000682
480 mm
Case of 5 for $55.08
Only null left
Add to Cart
 
17-623-852
View Documents
3M™
7000045911
530 mm
Case of 5 for $67.83
Only null left
Add to Cart
 
17-623-882
View Documents
3M™
7000120643
610 mm
Case of 5 for $100.98
Only null left
Add to Cart
 
Description

Description

  • Use to light scrub without dulling the finish
  • Can be used for everyday cleaning under and automatic scrubber
  • Also designed for spray buffing
  • Cleans when damp; buffs when dry
  • Use on rotary or automatic floor machines
  • *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
  • *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial and mid-life performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
Specifications

Specifications

Cleaning Pad
Buffing, Cleaning
Non-Woven Polyester Fiber
5/Cs.
Red
Rotary or Automatic Floor Machines
5100
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.