Learn More
Innovative Research Inc Pooled Human Red Blood Cells Lyophilized 1gm
Supplier:  Innovative Research Inc IWB3LY1GM
Encompass_Preferred
Pooled Human Red Blood Cells Lyophilized from Innovative Research are prepared from human whole blood that has been collected at an FDA-licensed facility located in the US. The red blood cells are separated from a solution of 1% dextran in whole blood via centrifugation and are not washed prior to lyophilization. A starting volume of approximately 5ml of red blood cells is used to create 1gm of lyophilized RBCs. 
Catalog No. 50-203-6201
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.