Learn More
Sartorius IncuCyte pHrodo Red E. coli Bioparticles for Phagocytosis
Catalog No. NC1612549 Shop All Sartorius Products
Encompass
One vial of 2 mg sterile lyophilized solid sufficient for 100-200 live-cell imaging phagocytosis tests in 96-well format
Catalog No. NC1612549 Supplier Sartorius Supplier No. 4615
One vial of 2 mg sterile lyophilized solid sufficient for 100-200 live-cell imaging phagocytosis tests in 96-well format