Learn More
Montana Molecular Red Fluorescent ACE2, C1100R, 5mL, CMV
SDP
Supplier:  Montana Molecular C1100R005
Encompass_Preferred
Express a red fluorescent ACE2 in your cells of interest. Use to investigate ACE2 inhibitors or facilitate cellular entry of Montana Molecular's Pseudo-SARS-CoV-2 assay. This assay is genetically encoded and packaged in BacMam for efficient transduction in most cell types - including uniform and near 100% efficiency in HEK293 cells as well as expression in primary or iPSC-derived cells.
Catalog No. NC1853240
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.