Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Red-Four (TM) Buffer Solution, pH 4.01 +/- 0.01 at 25°C, Color-coded RED, Reference Standard, Spectrum™ Chemical
SDP

Catalog No. 18606059 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
6 x 500 mL
20 L
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-059 20 L Poly Bottle
18-606-056 500 mL Poly Bottle
18-606-060 1 L Poly Bottle
18-606-057 6 x 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-059 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. B116320LT
Add to Cart
Add to Cart

B1163

Spectrum™ Chemical Red-Four (TM) Buffer Solution, pH 4.01 ± 0.01 at 25°C, Color-coded RED, Reference Standard

Specifications

CAS 877-24-7, 915-67-3, 7732-18-5
CAS Max % 0.01, 0.98
For Use With (Application) Calibration
For Use With (Equipment) pH Meter and Electrode
pH 4.01 ±0.01
Physical Form Aqueous Solution
Quantity 20 L
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.