Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Red Lead Solution ASTM D 49-83, Reagents
Red Lead Solution ASTM D 49-83, ASTM, for general laboratory use. Manufactured in ISO 9001 facility.
$2440.00
Specifications
Chemical Name or Material | Red Lead Solution ASTM D 49-83 |
---|---|
Color | Colorless to Light Yellow |
CAS | 7732-18-5, 6131-90-4, 64-19-7 |
Boiling Point | 100°C |
Grade | ASTM |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
01-334-073
|
Reagents Holdings Llc
CR01500020F |
20 L |
Each of 1 for $2,440.00
|
|
|||||
Specifications
Red Lead Solution ASTM D 49-83 | |
7732-18-5, 6131-90-4, 64-19-7 | |
ASTM | |
1.1033 | |
Non-sterile |
Colorless to Light Yellow | |
100°C | |
Cubitainer | |
20 L | |
Liquid |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title