Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Red Litmus pH Test Paper: Base Indicator
SCI_ED

Catalog No. S80846 Shop All Eisco Products
Change view
Click to view available options
pH Range:
6.8 to 8.1
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S80846 6.8 to 8.1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S80846 Supplier Eisco™ Supplier No. LITMUSREDPK12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Identifies a base or alkaline material with a pH of 6.8-8.1

  • Red Litmus Paper pH Tests
  • Indicates a basic solution or alkaline material
  • pH 6.8-8.1
  • 12 vials/pack; 100 strips/vial

Specifications

Color Indicator Red
pH Range 6.8 to 8.1
For Use With (Application) Detection of a Base or Alkaline Material
Quantity Pk. of 12 vials
Type Litmus Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.