Learn More
Andwin Scientific RED PRES VAC TUB 1/4IDX1/2 OD
SDP
Supplier:  Andwin Scientific 62995059
Encompass_Preferred
The Red Vacuum Rubber Tubing is ideal for vacuum work. It features a tensile strength of 20002600psi and durometer hardness of Shore A, 50. This product has a coil length of 50 feet and a wall thickness of 1/8".
Catalog No. NC9643412
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.